Order Entry
Czech Republic
ContactUsLinkComponent
L-Methionine ≥98.5% USP
L-Methionine ≥98.5% USP
Supplier:  Spectrum Chemical
Certificates
L-Methionine ≥98.5% USP
Supplier:  Spectrum Chemical
Certificates

Specification Test Results

  • Assay (Dried Basis):
    98.5 - 101.5%
  • Specific Rotation:
    +22.4 to +24.7°
  • pH (1 in 100):
    5.6 - 6.1
  • Loss On Drying:
    max. 0.3 %
  • Related Compounds:
    To Pass Test

Specification Test Results

Related Information

Frequently Bought Together