Order Entry
Czech Republic
ContactUsLinkComponent
Benzalkonium chloride solution 50% NF
Corrosive
Irritant
Environment
Benzalkonium chloride solution 50% NF
Supplier:  Spectrum Chemical
Certificates
Corrosive
Irritant
Environment
Benzalkonium chloride solution 50% NF
Supplier:  Spectrum Chemical

Specification Test Results

  • Assay (%):
    47.5

Specification Test Results

Frequently Bought Together