Order Entry
Czech Republic
ContactUsLinkComponent
Ammonium sulphide solution ACS
Ammonium sulphide solution ACS
Catalog # SPCMA1404-100ML
Supplier:  Spectrum Chemical
CAS Number:  
Ammonium sulphide solution ACS
Catalog # SPCMA1404-100ML
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :68.14 g/mol
Density:0,997
Storage Temperature:Ambient
UN:2683
ADR:86.13,II
CAS Number:12135-76-1
Formula:H₈N₂S
MDL Number:MFCD00010892