Order Entry
Czech Republic
ContactUsLinkComponent
N-Ethyldiisopropylamine ≥99%
Flammable
Corrosive
Toxic
Irritant
N-Ethyldiisopropylamine ≥99%
Catalog # SPCMD2163-200LTBL
Supplier:  Spectrum Chemical
CAS Number:  
Flammable
Corrosive
Toxic
Irritant
N-Ethyldiisopropylamine ≥99%
Catalog # SPCMD2163-200LTBL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pk:
    200 L
  • Seq:
    5549

Specifications

Specification Test Results

  • Assay:
    0.99

Specification Test Results

About this item

 :
 :129.25 g/mol
Density:0,742
Storage Temperature:Ambient
UN:2733
ADR:38,II
 :GHS02
 :GHS05
 :GHS06
 :GHS07
CAS Number:7087-68-5
Formula:C₈H₁₉N
MDL Number:MFCD00008868