Dimethyl succinate ≥99%
Dimethyl Succinate is a perfumery ingredient, also known as methyl butanedioate.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
: DBE-4 dibasic ester, Succinic acid dimethyl ester, 1,4-Butanedioic acid dimethyl ester
- :106-65-0
- :C₆H₁₀O₄
- :MFCD00008466
- :146.14 g/mol
- :1,117
- :Ambient
- Assay:99% min.
- Color (APHA):max.50
- Water:max. 0.1 %



