Order Entry
Czech Republic
ContactUsLinkComponent
Nicotinic acid (Niacin) ≥99.5%, powder FCC
Irritant
Nicotinic acid (Niacin) ≥99.5%, powder FCC
Supplier:  Spectrum Chemical
Certificates
Irritant
Nicotinic acid (Niacin) ≥99.5%, powder FCC
Supplier:  Spectrum Chemical
Certificates

Specification Test Results

  • Assay (Dried Basis):
    99.5-101.0%
  • Melting Range:
    234 - 238 °C
  • Loss On Drying:
    max. 1.0 %
  • Residue On Ignition:
    max. 0.1 %
  • Identification:
    To Pass Test

Specification Test Results

Related Information

Frequently Bought Together