Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Triammonium citrate

Triammonium citrate

Catalog #
Triammonium citrate
Catalog #
Supplier:  
CAS Number:  3458-72-8
Triammonium citrate
CAS Number:  3458-72-8
Product Details & Documents
Synonyms:  2-Hydroxy-1,2,3-propanetricarboxylic acid triammonium salt, Ammonium citrate tribasic, Citric acid triammonium salt, Triammonium 2-hydroxypropane-1,2,3-tricarboxylate
  • CAS Number:3458-72-8
  • Formula:C₆H₁₇N₃O₇
  • MDL Number:MFCD00036406
  • Molecular Weight:243.22 g/mol
  • Storage Temperature:Ambient