Orthophosphoric acid 85% ≥85% FCC
: Phosphoric acid
:7664-38-2
:
:H₃PO₄
:MFCD00011340
:158
:21
:1,685
:98 g/mol
:1805
:8 III
:14,07344
:GHS05
:GHS07
:Ambient
- Environmentally Preferable:
- Assay:85.0% min.
- Arsenic (As):max.3 mg/kg
- Cadmium (Cd):max. 3 mg/kg
- Fluoride:max. 10 mg/kg
- Lead (Pb):max. 3 mg/kg
Frequently Bought Together