Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-MPP7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-13737R-A647
 :  Bioss
Anti-MPP7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-13737R-A647
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    MPP7
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Epitope:
    301-400/576
  • Form:
    Liquid
  • Antigen synonyms:
    FLJ32798|MAGUK p55 subfamily member 7|Palmitoylated membrane protein 7|Membrane protein palmitoylated 7 MAGUK p55 subfamily member 7
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molecular weight:
    65 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human MPP7
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

The MAGUK (membrane-associated guanylate kinase homologs) family of proteins contain multiple protein-binding domains and are involved in cell junction organization, tumor suppression, and signaling. The MAGUK family is divided into four subfamilies: DLG-like, ZO1-like, p55-like and LIN2-like. MPP7 (MAGUK p55 subfamily member 7), also known as palmitoylated membrane protein 7, is a 576 amino acid protein that belongs to the MAGUK family and assists in the assembly of protein complexes. A peripheral membrane protein that exists as a heterodimer, MPP7 enhances tight junction formation and epithelial cell polarity. MPP7 contains one SH3 domain, a guanylate kinase-like domain, two L27 domains and a single PDZ (DHR) domain. The gene encoding MPP7 maps to human chromosome 10p11.23

Type: Primary
Antigen: MPP7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 647
Public Immunogen Range: 301-400/576
Host: Rabbit
Isotype: IgG
Reactivity: