Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Cadherin 20 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-Cadherin 20 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-13716R-A647
Supplier:  Bioss
Anti-Cadherin 20 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-13716R-A647
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cadherin 20
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Epitope:
    301-400/801
  • Form:
    Liquid
  • Antigen synonyms:
    FLJ37047|CDH7L3|Cadherin 20, type 2|Cdh7|Cadherin 20|cadherin-20
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molecular weight:
    82 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Cadherin 20
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

The cadherins are a family of Ca2+-dependent adhesion molecules that function to mediate cell-cell binding critical to the maintenance of structure and morphogenesis. Cadherins each contain a large extracellular domain at the N-terminus, which is characterized by a series of five homologous repeats, the most distal of which is thought to be responsible for binding specificity. Cadherin-20, also known as protocadherin gamma B4, is a member of the protocadherin gamma gene cluster. Expressed primariy in fibroblasts, cadherin-20 plays a critical role in neuronal cell-cell connections in the brain. Through its ability to mediate cell-cell adhesion, caderin-20 is thought to help the body respond to injury by regulating post-traumatic wound healing.

Type: Primary
Antigen: Cadherin 20
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 647
Public Immunogen Range: 301-400/801
Host: Rabbit
Isotype: IgG
Reactivity: