- Antibody type:Primary
- Antigen name:FNBP3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:851-957/957
- Form:Liquid
- Gene ID:55660
- Antigen synonyms:NY REN 6 antigen|PR40A_HUMAN|HYPA|Pre mRNA processing factor 40 homolog A|FNBP 3|Huntingtin yeast partner A|Formin-binding protein 11|PRPF40A|PRP40 pre mRNA processing factor 40 homolog A S cerevisiae|Huntingtin interacting protein HYPA|Renal carcinoma antigen NY-REN-6|FLAF 1|FBP 11|FLAF1|Formin-binding protein 3|HIP-10|Fas ligand-associated factor 1|Fas ligand associated factor 1|FBP11|PRP40 pre mRNA processing factor 40 homolog A|NY REN 6|PRPF 40A|Huntingtin interacting protein A|HIP10|PRP40 pre mRNA processing factor 40 homolog A yeast|Formin binding protein 11|Huntingtin-interacting protein A|Pre-mRNA-processing factor 40 homolog A|Formin binding protein 3|Huntingtin-interacting protein 10
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:109 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human FNBP3
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Fas ligand associated factor 1; Fas ligand-associated factor 1; FBP 11; FBP11; FLAF 1; FLAF1; FNBP 3; Formin binding protein 11; Formin binding protein 3; Formin-binding protein 11; Formin-binding protein 3; HIP-10; HIP10; Huntingtin interacting protein A; Huntingtin interacting protein HYPA; Huntingtin yeast partner A; Huntingtin-interacting protein 10; Huntingtin-interacting protein A; HYPA; NY REN 6; NY REN 6 antigen; PR40A_HUMAN; Pre mRNA processing factor 40 homolog A; Pre-mRNA-processing factor 40 homolog A; PRP40 pre mRNA processing factor 40 homolog A (S cerevisiae); PRP40 pre mRNA processing factor 40 homolog A (yeast); PRP40 pre mRNA processing factor 40 homolog A; PRPF 40A; PRPF40A; Renal carcinoma antigen NY-REN-6.
Type: Primary
Antigen: FNBP3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 851-957/957
Host: Rabbit
Isotype: IgG
Reactivity: Human