- Antibody type:Primary
- Antigen name:COPS2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:51-185/443
- Form:Liquid
- Gene ID:1447
- Antigen synonyms:Signalosome subunit 2|ALIEN|COP9 signalosome complex subunit 2|SGN2|Thyroid receptor-interacting protein 15|SGN 2|Cops2|CSN2|COPS 2|COP9 constitutive photomorphogenic homolog subunit 2|Thyroid Hormone Receptor Interactor15|JAB1-containing signalosome subunit 2|TRIP 15|Thyroid Hormone Receptor Interactor 15|TRIP-15|COP9|Alien homolog|JAB1 containing signalosome subunit 2|CSN2_HUMAN|COP 9|TR-interacting protein 15
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human COPS2/TRIP15
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Essential component of the COP9 signalosome complex (CSN), a complex involved in various cellular and developmental processes. The CSN complex is an essential regulator of the ubiquitin (Ubl) conjugation pathway by mediating the deneddylation of the cullin subunits of SCF-type E3 ligase complexes, leading to decrease the Ubl ligase activity of SCF-type complexes such as SCF, CSA or DDB2. The complex is also involved in phosphorylation of p53/TP53, c-jun/JUN, IkappaBalpha/NFKBIA, ITPK1 and IRF8/ICSBP, possibly via its association with CK2 and PKD kinases. CSN-dependent phosphorylation of TP53 and JUN promotes and protects degradation by the Ubl system, respectively. Involved in early stage of neuronal differentiation via its interaction with NIF3L1.
Type: Primary
Antigen: COPS2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-185/443
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat