Anti-GTF2H1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-8175R-A750
: Bioss
- Antibody type:Primary
- Antigen name:GTF2H1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:201-300/548
- Form:Liquid
- Gene ID:2965
- Antigen synonyms:Basic transcription factor 2 62 kDa subunit|P62|BTF2|TFB1|BTF2 p62|General transcription factor IIH polypeptide 1|GTF2H1|General transcription factor IIH subunit 1|TFIIH|TFIIH basal transcription factor complex p62 subunit
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:62 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GTF2H1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Component of the core-TFIIH basal transcription factor involved in nucleotide excision repair (NER) of DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II.
Type: Primary
Antigen: GTF2H1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/548
Host: Rabbit
Isotype: IgG
Reactivity: Human