Specifications
- Antibody type:Primary
- Antigen name:CCDC34
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Epitope:177-280/373
- Form:Liquid
- Gene ID:91057
- Antigen synonyms:CCD34_HUMAN|Ccdc34|RAMA3|Coiled coil domain containing protein 34|L15|Renal carcinoma antigen NY REN 41
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:43 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human CCDC34
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
The coiled-coil domain is a structural motif found in proteins that are involved in a diverse array of biological functions such as the regulation of gene expression, cell division, membrane fusion and drug extrusion and delivery. CCDC34 (coiled-coil domain containing 34), also known as Renal carcinoma antigen NY-REN-41, is a 373 amino acid protein that is expressed in testis, breast, lung, placenta, liver and small intestine. A translocation between the short arms of chromosomes 11 and 18 affecting the CCDC34 gene has been identified in a patient with hamartoma of the retinal pigment epithelium. There are two isoforms of CCDC34 that are produced as a result of alternative splicing events.
Type: Primary
Antigen: CCDC34
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 177-280/373
Host: Rabbit
Isotype: IgG
Reactivity: Rat