Anti-CDC2L6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-7070R-A750
: Bioss
- Antibody type:Primary
- Antigen name:CDC2L6
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Epitope:21-120/502
- Form:Liquid
- Gene ID:23097
- Antigen synonyms:CDC2 related protein kinase 6|Cell division cycle 2-like protein kinase 6|Cyclin dependent kinase CDC2 like 11|Death-preventing kinase|Cyclin-dependent kinase 11|CDC2-related protein kinase 6|Cyclin dependent kinase 11|CDK8 like cyclin dependent kinase|Cell division cycle 2 like 6 CDK8 like|CDK11|Cell division cycle 2 like protein kinase 6|Cyclin-dependent kinase 19|CDK19_HUMAN|Death preventing kinase|Cell division protein kinase 19|Cdk19
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:57 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human CDC2L6
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
This gene encodes a protein that is one of the components of the Mediator coactivator complex. The Mediator complex is a multiprotein complex required for transcriptional activation by DNA binding transcription factors of genes transcribed by RNA polymerase II. The protein encoded by this gene is similar to cyclin-dependent kinase 8 which can also be a component of the Mediator complex.
Type: Primary
Antigen: CDC2L6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-120/502
Host: Rabbit
Isotype: IgG
Reactivity: Mouse