Order Entry
Czech Republic
ContactUsLinkComponent
Anti-PPAPDC3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-PPAPDC3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6918R-A680
Supplier:  Bioss
Anti-PPAPDC3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6918R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    PPAPDC3
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    121-220/271
  • Form:
    Liquid
  • Gene ID:
    84814
  • Antigen synonyms:
    C9orf67|Ppapdc3|PPAC3_HUMAN|Phosphatidic acid phosphatase type 2 domain-containing protein 3|Probable lipid phosphate phosphatase PPAPDC3
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    30 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human PPAPDC3
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Plays a role as negative regulator of myoblast differentiation, in part through effects on MTOR signaling. Has no detectable enzymatic activity.PPAPDC3, also known as nuclear envelope transmembrane protein 39 (NET39), was initially discovered in an in silico screen for secreted or membrane proteins. It is a member of the PAP2 superfamily of phosphatases and haloperoxidases. PPAPDC3 has recently been shown to act as a negative regulator of myoblast differentiation by diminishing the activity of the mammalian target of rapamycin TOR. PPAPDC3 is highly expressed in cardiac and skeletal muscle and becomes strongly upregulated during cultured myoblast differentiation tissues. Overexpression of PPAPDC3 in myoblasts repressed myogenesis while knockdown by RNA interference promoted differentiation indicating its part in the regulatory mechanism for myogenesis.

Type: Primary
Antigen: PPAPDC3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 121-220/271
Host: Rabbit
Isotype: IgG
Reactivity: