Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-NIRF Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6389R-A680
Supplier:  Bioss
Anti-NIRF Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6389R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    NIRF
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Epitope:
    15-100/802
  • Form:
    Liquid
  • Gene ID:
    115426
  • Antigen synonyms:
    Nuclear zinc finger protein Np97|Np95 like RING finger protein|E3 ubiquitin-protein ligase UHRF2|Np95-like ring finger protein|Np95/ICBP90 like RING finger protein|Ubiquitin-like-containing PHD and RING finger domains protein 2|UHRF2_HUMAN|RP11-472F14.2|RING finger protein 107|Np95/ICBP90-like RING finger protein|Ubiquitin like containing PHD and RING finger domains protein 2|RNF 107|Uhrf2|Nuclear protein 97|Ubiquitin-like PHD and RING finger domain-containing protein 2
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    90 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human NIRF/RNF107
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

E3 ubiquitin-protein ligase which mediates ubiquitination and subsequent proteasomal degradation of PCNP. May participate in methylation-dependent transcriptional regulation. Important for G1/S transition. Overexpression causes G1 phase cell arrest.

Type: Primary
Antigen: NIRF
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 15-100/802
Host: Rabbit
Isotype: IgG
Reactivity: