Order Entry
Czech Republic
ContactUsLinkComponent
Anti-LMO4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-LMO4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-5966R-A680
Supplier:  Bioss
Anti-LMO4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-5966R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    LMO4
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    65-165/165
  • Form:
    Liquid
  • Gene ID:
    8543
  • Antigen synonyms:
    LIM domain transcription factor LMO 4|LMO4|LIM domain transcription factor LMO4|LIM domain only 4|Breast tumor autoantigen|LMO 4|LMO-4|LMO4_HUMAN|LIM domain only protein 4
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    18 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human LMO4
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

The LIM-only (LMO) proteins, LMO1 and LMO2, are nuclear factors that are characterised by a conserved LIM domain (1). The LIM domain consists of a cysteine-rich zinc-binding motif that is present in a variety of transcription factors, including the LIM homeobox (LHX) proteins expressed in the central nervous system and involved in cell differentiation (2). LMO1 and LMO2 are expressed in the adult CNS in a cell type-specific manner, where they are differentially regulated by neuronal activity and are involved in regulating the cellular differentiated phenotype of neurons (3). LMO2 lacks a specific DNA-binding homeobox domain but rather assembles into transcriptional regulatory complexes to mediate gene expression by interacting with the widely expressed nuclear LIM interactor (NLI) (4). NLI, known also as CLIM-1, and the related protein CLIM-2 facilitate the formation of heteromeric LIM complexes and also enhance the nuclear retention of LIM proteins (5). LMO2 and the related protein LMO4 are expressed in thymic precursor cells (6). LMO4 is also expressed in mature T cells, cranial neural crest cells, somite, dorsal limb bud mesenchyme, motor neurons, and Schwann cell progenitors (7).

Type: Primary
Antigen: LMO4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 65-165/165
Host: Rabbit
Isotype: IgG
Reactivity: