Anti-ADAM19 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-5850R-A680
: Bioss
- Antibody type:Primary
- Antigen name:ADAM19
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:251-350/955
- Form:Liquid
- Gene ID:8728
- Antigen synonyms:MLTNB|FKSG34|Disintegrin and metalloproteinase domain-containing protein 19|Meltrin-beta|ADAM19|Metalloprotease and disintegrin dendritic antigen marker|MADDAM|ADAM 19
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:82 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human ADAM19
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Participates in the proteolytic processing of beta-type neuregulin isoforms which are involved in neurogenesis and synaptogenesis, suggesting a regulatory role in glial cell. Also cleaves alpha-2 macroglobulin. May be involved in osteoblast differentiation and/or osteoblast activity in bone (By similarity).
Type: Primary
Antigen: ADAM19
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 251-350/955
Host: Rabbit
Isotype: IgG
Reactivity: Human