Zadání objednávky
Czech Republic
ContactUsLinkComponent
 
Anti-IL2 Receptor beta Tyr364 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-11300R-A680
Supplier:  Bioss
Anti-IL2 Receptor beta Tyr364 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-11300R-A680
Supplier:  Bioss

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    IL2 Receptor beta Tyr364
  • Klonovalita:
    Polyclonal
  • Spojení:
    Alexa Fluor® 680
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG
  • Western blot:
    Yes
  • Formulář:
    Liquid
  • Identifikační číslo genu:
    3560
  • Synonyma antigenu:
    High affinity IL-2 receptor subunit beta|IL-2RB|p75|IL-2R subunit beta|Interleukin-2 receptor subunit beta|p70-75|IL2RB|IL-2 receptor subunit beta
  • Modifikace:
    Phosphorylation (Tyr364)
  • Skladovací pufr:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molekulární hmotnost:
    58 kDa
  • Skladovací teplota:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentrace:
    1 µg/µl
  • Transportní teplota:
    4 °C
  • Imunogen:
    KLH conjugated synthetic phosphopeptide derived from human IL2 Receptor beta around the phosphorylation site of Tyr364
  • Testované aplikace:
    ICC
  • Čištění:
    Purified by Protein A
  • Typ balení:
    Vial
  • Bal.:
    100 µl

Specifications

About this item

IL2 Receptor beta (CD122) is a member of the immunoglobulin superfamily that forms the high affinity IL2 receptor with CD25 and CD132. This receptor chain, which is also shared by the IL15 receptor, is constitutively expressed by NK cells and at lower levels by T cells, B cells, monocytes, and macrophages. The IL2 Receptor beta chain can combine with either the common gamma subunit (gamma c) alone or the gamma c subunit and the IL2 Receptor alpha subunit to generate intermediate or high affinity IL2 receptor complexes, respectively. CD122 levels can be upregulated by activation stimuli such as IL2.

Type: Primary
Antigen: IL2 Receptor beta Tyr364
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: