Order Entry
Czech Republic
ContactUsLinkComponent
Anti-HSP 40-4/HDJ2/DNAJA1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-HSP 40-4/HDJ2/DNAJA1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-11277R-A680
Supplier:  Bioss
Anti-HSP 40-4/HDJ2/DNAJA1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-11277R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HSP 40-4/HDJ2/DNAJA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    21-110/397
  • Form:
    Liquid
  • Antigen synonyms:
    OTTHUMP00000021193|HDJ-2|DJ 2|DNAJ2|Heat shock 40 kDa protein 4|heat shock protein DNAJ like 2|HSJ-2|Heat shock protein J2|DnaJ Hsp40 homolog, subfamily A, member 1|HSPF4|DNJA1_HUMAN|DjA1|HDJ2|Neural precursor cell expressed developmentally down regulated 7|DJ2|HSDJ|NEDD7|HSJ2|hDJ 2|DnaJ protein homolog 2|Human DnaJ protein 2|DnaJ homolog subfamily A member 1|Dnaja1
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    45 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human HSP 40-4
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

DnaJ-like proteins interact with HSP 70 molecular chaperones and function to facilitate protein folding and mitochondrial protein import. HSP 40-4, also known as HDJ2, is the human DnaJ homolog that functions as a co-chaperone with a cysteine-rich zinc finger domain. The cellular redox enzyme thioredoxin interacts with HSP 40-4, and oxidation and reduction reversibly regulate HSP 40-4 function in response to the changing redox states of the cell. The zinc finger domain of HSP 40-4 may act as a redox sensor of chaperone-mediated protein-folding machinery, since HSP 40-4 inactivation leads to the oxidation of cysteine thiols and a simultaneous release of coordinated zinc. Loss of the HSP 40-4 protein may be linked to severe defects in spermatogenesis that involve aberrant androgen Signalling.

Type: Primary
Antigen: HSP 40-4/HDJ2/DNAJA1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 21-110/397
Host: Rabbit
Isotype: IgG
Reactivity: Mouse