Specifications
- Antibody type:Primary
- Antigen name:SPON2/M spondin/Mindin
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:21-120/331
- Form:Liquid
- Antigen synonyms:Dferentially expressed in cancerous and non-cancerous lung cells 1|SPON2_HUMAN|DIL-1|Mindin|Spondin 2|DIL1|Spondin-2|SPON 2|M spondin|DIL 1|Spon2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:33 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human SPON2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
The Thrombospondin proteins, Thrombospondins 1-4 and Thrombospondin 5 (also designated COMP), compose a family of glycoproteins that are involved in cell-to-cell and cell-to-matrix Signalling. These extracellular, cell-surface proteins form complexes of both homo- and heteromultimers. Spondin-2, or Mindin, is also designated DIL-1 for its differential expression in cancerous and non-cancerous lung cells. Full-length SPON2 cDNA encodes a 331 amino acid protein with a domain arrangement similar to zebrafish F-Spondin and Mindin-1/Mindin-2: an FS1 domain, an FS2 domain, a hydrophobic signal sequence in the N-terminus and a Thrombospondin type I repeat. Immunoblot analysis demonstrates expression of dimers and oligomers in a concentration-dependent manner under nonreducing conditions.
Type: Primary
Antigen: SPON2/M spondin/Mindin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-120/331
Host: Rabbit
Isotype: IgG
Reactivity: