Anti-protein 4.2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-9879R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:protein 4.2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:201-300/691
- Form:Liquid
- Gene ID:2038
- Antigen synonyms:PA|EPB42_HUMAN|Erythrocyte membrane protein band 4.2|protein 4.2|MGC116735|Erythrocyte surface protein band 4.2|SPH5|Erythrocyte protein 4.2|EPB42|protein4.2|P4.2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human EPB42/protein 4.2
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Protein 4.2; protein4.2; EPB42; EPB42_HUMAN; Erythrocyte membrane protein band 4.2; Erythrocyte protein 4.2; Erythrocyte surface protein band 4.2; MGC116735; P4.2; PA; SPH5.
Type: Primary
Antigen: protein 4.2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/691
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat