Anti-NOL1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-9710R-A750
: Bioss
- Antibody type:Primary
- Antigen name:NOL1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:1-100/812
- Form:Liquid
- Gene ID:4839
- Antigen synonyms:proliferation Associated Nucleolar Antigen: NOL 1|NUCLEOLAR ANTIGEN p120|NOP2_HUMAN
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human NOL1
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Proliferation-associated nucleolar antigen is expressed in mid G1 phase with peak level during the S phase and a rapid degradation during late mitosis. Its expression in breast carcinoma is correlated with patient prognosis.
Type: Primary
Antigen: NOL1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/812
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat