Specifications
- Antibody type:Primary
- Antigen name:C20orf11
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:1-100/228
- Form:Liquid
- Antigen synonyms:Twa1|Two hybrid associated protein 1|Two hybrid-associated protein 1 with Ran BP-M|CT011_HUMAN|Protein C20orf11|Chromosome 20 open reading frame 11|Two hybrid associated protein 1 with RanBPM|Two hybrid-associated protein 1 with RanBPM
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human TWA1/C20orf11
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
TWA1 is a 228 amino acid nuclear protein that is highly conserved throughout evolution. TWA1 interacts with Ran BP-M, a protein that is implicated in a diverse array of cellular processes including DNA replication, entry into and exit from mitosis, and the transport of RNA and proteins through the nuclear pore complex. Together, Ran BP-M and TWA1 form a complex with cytoplasmic Muskelin, a mediator of cell spreading. It is thought that this complex plays a role in the Ran GTPase cycle and therefore a potential role in the cell cycle. TWA1 contains a LisH-CTLH motif which is usually found in proteins that are involved in nucleokinesis, chromosome segregation, cell migration and microtubule dynamics. The gene encoding TWA1 maps to human chromosome 20, which comprises approximately 2% of the human genome and contains nearly 63 million bases that encode over 600 genes.
Type: Primary
Antigen: C20orf11
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/228
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat