Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SPA-1/SIPA-1 Rabbit Polyclonal Antibody (HRP (Horseradish PE (Phycoerythrin)rOxidase))
Anti-SPA-1/SIPA-1 Rabbit Polyclonal Antibody (HRP (Horseradish PE (Phycoerythrin)rOxidase))
Catalog # BOSSBS-1602R-HRP
Supplier:  Bioss
Anti-SPA-1/SIPA-1 Rabbit Polyclonal Antibody (HRP (Horseradish PE (Phycoerythrin)rOxidase))
Catalog # BOSSBS-1602R-HRP
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SPA-1/SIPA-1
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Epitope:
    351-450/1042
  • Form:
    Liquid
  • Gene ID:
    6494
  • Antigen synonyms:
    SIPA1_HUMAN|MGC102688|Signal induced proliferation associated 1|GTPase activating protein Spa 1|Sipa1|MGC17037|Signal induced proliferation associated protein 1|Spa1.|GTPase-activating protein Spa-1|signal-induced proliferation-associated gene 1|Sipa-1|SIPA 1|Signal induced proliferation associated gene 1|p130 SPA1|p130 SPA-1|Signal-induced proliferation-associated protein 1
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    112 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human SPA-1
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

The product of this gene is a mitogen induced GTPase activating protein (GAP). It exhibits a specific GAP activity for Ras-related regulatory proteins Rap1 and Rap2, but not for Ran or other small GTPases. This protein may also hamper mitogen-induced cell cycle progression when abnormally or prematurely expressed. It is localised to the perinuclear region. Two alternatively spliced variants encoding the same isoform have been characterised to date.

Type: Primary
Antigen: SPA-1/SIPA-1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 351-450/1042
Host: Rabbit
Isotype: IgG
Reactivity: