Anti-SPA-1/SIPA-1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-1602R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:SPA-1/SIPA-1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:351-450/1042
- Form:Liquid
- Gene ID:6494
- Antigen synonyms:SIPA1_HUMAN|MGC102688|Signal induced proliferation associated 1|GTPase activating protein Spa 1|Sipa1|MGC17037|Signal induced proliferation associated protein 1|Spa1.|GTPase-activating protein Spa-1|signal-induced proliferation-associated gene 1|Sipa-1|SIPA 1|Signal induced proliferation associated gene 1|p130 SPA1|p130 SPA-1|Signal-induced proliferation-associated protein 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:112 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human SPA-1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
The product of this gene is a mitogen induced GTPase activating protein (GAP). It exhibits a specific GAP activity for Ras-related regulatory proteins Rap1 and Rap2, but not for Ran or other small GTPases. This protein may also hamper mitogen-induced cell cycle progression when abnormally or prematurely expressed. It is localised to the perinuclear region. Two alternatively spliced variants encoding the same isoform have been characterised to date.
Type: Primary
Antigen: SPA-1/SIPA-1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 350
Public Immunogen Range: 351-450/1042
Host: Rabbit
Isotype: IgG
Reactivity: