Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Ghrelin 28 Rabbit Polyclonal Antibody (Cy5®)
Anti-Ghrelin 28 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-1375R-CY5
Supplier:  Bioss
Anti-Ghrelin 28 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-1375R-CY5
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ghrelin 28
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Epitope:
    24-51/117
  • Form:
    Liquid
  • Gene ID:
    58991
  • Antigen synonyms:
    motilin-related peptide|grehlin|growth hormone secretagogue|protein M46|growth hormone-releasing peptide|Appetite-regulating hormone
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    13 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide of mouse Ghrelin
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Ghrelin is an endogenous ligand for the growth hormone secretagogue receptor and is involved in regulating growth hormone release. Ghrelin is derived from a preprohormone called preproghrelin, which also generates a second peptide called obestatin. Obestatin is an endogenous ligand for the orphan G protein-coupled receptor GPR39 and is involved in satiety and decreased food intake. Also known as Appetite regulating hormone; GHRL; Growth hormone releasing peptide; Growth hormone secretagogue; M46 protein; Motilin related peptide; MTLRP; Obestatin; Obestatin preprohormone; PRO1066; UNQ524. Sequence notes: Gly-Ser-Ser-Phe-Leu-Ser-Pro- Glu-His-Gln-Lys-Ala-Gln-Gln-Arg-Lys-Glu- Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro- Arg (mo, rat).

Type: Primary
Antigen: Ghrelin 28
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 24-51/117
Host: Rabbit
Isotype: IgG
Reactivity: Rat