Anti-Tenascin C Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-1327R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Tenascin C
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:2101-2201/2201
- Form:Liquid
- Gene ID:3371
- Antigen synonyms:TN C|HXB|GP 150 225|Tenascin-C.|Neuronectin|Glioma associated extracellular matrix antigen|Cytotactin|Myotendinous antigen|JI|Hexabrachion|TN|TNC.Tn-C|GMEM|TenascinC|Tenascin C
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:230 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Tenascin C
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Tenascin, also known as hexabrachion and cytotactin, is an extracellular matrix protein with a spatially and temporally restricted tissue distribution. It is a hexameric, multidomain protein with disulfide linked subunits of 190 to 240 kD, originally characterised as 'myotendinous antigen.' In the embryo it is present in dense mesenchyme surrounding developing epithelia and in developing cartilage and bone. In the adult, tenascin remains present in tendons and myotendinous junctions in the perichondrium and periosteum, as well as in smooth muscle.
Type: Primary
Antigen: Tenascin C
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 2101-2201/2201
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog