Specifications
- Antibody type:Primary
- Antigen name:Tyrosinase
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:155-250/529
- Form:Liquid
- Gene ID:7299
- Antigen synonyms:TYRO_HUMAN.|Tumor rejection antigen AB|tyrosinase oculocutaneous albinism IA|Oculocutaneous albinism IA|Monophenol monooxygenase|OCAIA|LB24 AB|OCA1A|TYR|LB24-AB|SK29-AB|SK29 AB
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:56 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Tyrosinase
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Tyrosinase is the key enzyme for melanin,synthesis in mammalian melanocytes and has been considered to be a unique marker for the study of melanocyte differentiation. A cDNA library was constructed from poly(A)+ mRNA from mouse melanocytes and screened using anti- tyrosinase antiserum and oligonucleotide probes corresponding to amino acid sequence of tyrosinase. sequencing of some cDNA clones positive in these screenings gave a nucleotide sequence of 1838 nucleotides including a open reading frame of 1344 nucleotides that was found to correspond exactly to the amino acid sequence of the cyanogen bromide fragments of tyrosinase.
Type: Primary
Antigen: Tyrosinase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 155-250/529
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat