Anti-AACT Alpha1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-0094R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:AACT Alpha1
- Antigen symbol:AACT Alpha1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:121-320/423
- Form:Liquid
- Gene ID:12
- Antigen synonyms:Cell growth-inhibiting gene 24/25 protein|Alpha-1-antichymotrypsin|Serpin A3|AACT|ACT|SERPINA3|GIG24|GIG25
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:45 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human AACT-Alpha1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Although its physiological function is unclear, it can inhibit neutrophil cathepsin G and mast cell chymase, both of which can convert angiotensin-1 to the active angiotensin-2.
Type: Primary
Antigen: AACT Alpha1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 121-320/423
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat