Order Entry
Czech Republic
ContactUsLinkComponent
Anti-beta Amyloid 1-42 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-beta Amyloid 1-42 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-0076R-A750
Supplier:  Bioss
Anti-beta Amyloid 1-42 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-0076R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    beta Amyloid 1-42
  • Antigen symbol:
    beta Amyloid 1-42
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Epitope:
    35-42/42
  • Form:
    Liquid
  • Gene ID:
    351
  • Antigen synonyms:
    APP|A4|PreA4|CTFgamma|CVAP|ABETA|Protease nexin-II|APPI|Beta-amyloid precursor protein|AD1|PN-II|ABPP|Alzheimer disease amyloid protein|Amyloid beta A4 protein|Cerebral vascular amyloid peptide|AAA|PN2|Amyloid precursor protein
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    4,3/75 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human beta-Amyloid(35-42)
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Functions as a cell surface receptor and performs physiological functions on the surface of neurons relevant to neurite growth, neuronal adhesion and axonogenesis. Involved in cell mobility and transcription regulation through protein-protein interactions. Can promote transcription activation through binding to APBB1-KAT5 and inhibits Notch signaling through interaction with Numb. Couples to apoptosis-inducing pathways such as those mediated by G(O) and JIP. Inhibits G(o) alpha ATPase activity (By similarity). Acts as a kinesin I membrane receptor, mediating the axonal transport of beta-secretase and presenilin 1. Involved in copper homeostasis/oxidative stress through copper ion reduction. In vitro, copper-metallated APP induces neuronal death directly or is potentiated through Cu(2+)-mediated low-density lipoprotein oxidation. Can regulate neurite outgrowth through binding to components of the extracellular matrix such as heparin and collagen I and IV. The splice isoforms that contain the BPTI domain possess protease inhibitor activity. Induces a AGER-dependent pathway that involves activation of p38 MAPK, resulting in internalisation of amyloid-beta peptide and leading to mitochondrial dysfunction in cultured cortical neurons. Provides Cu(2+) ions for GPC1 which are required for release of nitric oxide (NO) and subsequent degradation of the heparan sulfate chains on GPC1. Beta-amyloid peptides are lipophilic metal chelators with metal-reducing activity. Bind transient metals such as copper, zinc and iron. In vitro, can reduce Cu(2+) and Fe(3+) to Cu(+) and Fe(2+), respectively. Beta-amyloid 42 is a more effective reductant than beta-amyloid 40. Beta-amyloid peptides bind to lipoproteins and apolipoproteins E and J in the CSF and to HDL particles in plasma, inhibiting metal-catalysed oxidation of lipoproteins. Beta-APP42 may activate mononuclear phagocytes in the brain and elicit inflammatory responses.

Type: Primary
Antigen: beta Amyloid 1-42
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 35-42/42
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse