Anti-FAM13B1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-5757R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:FAM13B1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:841-915/915
- Form:Liquid
- Antigen synonyms:GAP-like protein N61|FAM13B1|Fam13b|KHCHP|FA13B_HUMAN|Protein FAM13B|Family with sequence similarity 13, member B1|N61|C5orf5
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:105 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human FAM13B1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
FAM13B1.
Type: Primary
Antigen: FAM13B1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 841-915/915
Host: Rabbit
Isotype: IgG
Reactivity: Human