Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-XPB Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-4260R-A680
 :  Bioss
Anti-XPB Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-4260R-A680
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    XPB
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Epitope:
    601-700/782
  • Form:
    Liquid
  • Gene ID:
    2071
  • Antigen synonyms:
    ERCC3|DNA repair protein complementing XP-B cells|GTF2H|XPBC|BTF2|DNA excision repair protein ERCC-3|TFIIH basal transcription factor complex helicase XPB subunit|Xeroderma pigmentosum group B-complementing protein|XPB|BTF2 p89|TFIIH|Basic transcription factor 2 89 kDa subunit|TFIIH p89|TFIIH 89 kDa subunit|TFIIH basal transcription factor complex 89 kDa subunit|RAD25
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    89 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human XPB/ERCC3
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

ATP-dependent 3'-5' DNA helicase, component of the core-TFIIH basal transcription factor, involved in nucleotide excision repair (NER) of DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II. Acts by opening DNA either around the RNA transcription start site or the DNA damage.

Type: Primary
Antigen: XPB
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 601-700/782
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat