Anti-S100-A9 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-2697R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:S100-A9
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Epitope:51-114/114
- Form:Liquid
- Gene ID:6280
- Antigen synonyms:CAGB|6B8AG|MRP14|Calprotectin L1H subunit|CFAG|Protein S1-A9|Leukocyte L1 complex heavy chain|S1 calcium-binding protein A9|NIF|MAC387|MIF|L1AG|CGLB|Calgranulin-B|P14|MRP-14|LIAG|S1A9|Migration inhibitory factor-related protein 14
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:14 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human S100-A9
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localised in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and altered expression of this protein is associated with the disease cystic fibrosis.
Type: Primary
Antigen: S100-A9
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-114/114
Host: Rabbit
Isotype: IgG
Reactivity: Human