Anti-CD32 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-2573R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:CD32
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:201-300/317
- Form:Liquid
- Gene ID:2212
- Antigen synonyms:CD32|IgG Fc receptor II-a|FcGR|CD32A|IGFR2|FCG2|FcRII-a|FCGR2A1|CDw32|Fc-gamma RII-a|FCGR2|FCGR2A|Low affinity immunoglobulin gamma Fc region receptor II-a|Fc-gamma-RIIa
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:31 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human CD32
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Binds to the Fc region of immunoglobulins gamma. Low affinity receptor. By binding to IgG it initiates cellular responses against pathogens and soluble antigens. Promotes phagocytosis of opsonized antigens.
Type: Primary
Antigen: CD32
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/317
Host: Rabbit
Isotype: IgG
Reactivity: Human