Anti-CCL19/MIP-3 beta Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-2454R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:CCL19/MIP-3 beta
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:26-108/108
- Form:Liquid
- Gene ID:24047
- Antigen synonyms:Epstein-Barr virus-induced molecule 1 ligand chemokine|MIP3B|Scya19|C-C motif chemokine 19|ELC|EBI1 ligand chemokine|exodus-3|Small-inducible cytokine A19|Ccl19|CKb11
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:9.2 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from mouse CCL19
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Strongly chemotactic for naive (L-selectinhi) CD4 T-cells and for CD8 T-cells and weakly attractive for resting B-cells and memory (L-selectinlo) CD4 T-cells. May play a role in promoting encounters between recirculating T-cells and dendritic cells and in the migration of activated B-cells into the T-zone of secondary lymphoid tissues. Binds to chemokine receptor CCR7. Binds to atypical chemokine receptor ACKR4 and mediates the recruitment of beta-arrestin (ARRB1/2) to ACKR4.
Type: Primary
Antigen: CCL19/MIP-3 beta
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 26-108/108
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat