Anti-BDKRB2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-2422R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:BDKRB2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:51-150/391
- Form:Liquid
- Gene ID:624
- Antigen synonyms:BKR2|B2 bradykinin receptor|BDKRB2|BK2|BRB2|BK-2 receptor|BK-2|B2R
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:43 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Bdkrb2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Receptor for bradykinin. It is associated with G proteins that activate a phosphatidylinositol-calcium second messenger system.
Type: Primary
Antigen: BDKRB2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/391
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat