Anti-IFIT3 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Catalog # BOSSBS-15515R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:IFIT3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:101-200/490
- Form:Liquid
- Antigen synonyms:Interferon-induced 60 kDa protein|CIG 49|IFIT-3|IFIT4|RP11-149I23,4|IFI60|Ifit3|RIG-G|IFIT-4|Retinoic acid-induced gene G protein|Interferon-induced protein with tetratricopeptide repeats 3|ISG-60|Interferon-induced protein with tetratricopeptide repeats 4|IRG2|IFIT3_HUMAN|Interferon induced protein with tetratricopeptide repeats 3|RIGG|GARG 49|Interferon induced 60 kDa protein|Retinoic acid induced gene G protein|RIG G|P60|ISG60|IFI-60K|CIG49|IFI60K
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:56 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human IFIT3
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
IFIT3.
Type: Primary
Antigen: IFIT3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/490
Host: Rabbit
Isotype: IgG
Reactivity: