Anti-C6ORF222 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
BOSSBS-15241R-A680
: Bioss
- Antibody type:Primary
- Antigen name:C6orf222
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:531-630/652
- Form:Liquid
- Gene ID:389384
- Antigen synonyms:Uncharacterized protein C6orf222|C6orf222
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:72 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human C6ORF222
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
C6orf222.
Type: Primary
Antigen: C6orf222
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 531-630/652
Host: Rabbit
Isotype: IgG
Reactivity: Human