Dulbecco's Modification of Eagle's Medium, MP Biomedicals
Catalog # ICNA091233454
Supplier: MP Biomedicals
CAS Number:
Specifications
- Media Format:Liquid
- Pk:500 mL
- Description:Dulbecco's Modified Eagle's Medium (DMEM) (1X Solution) with 20 mM HEPES, without L-glutamine or sodium bicarbonate
- Storage temperature:4 °C
Specifications
About this item
Dulbecco's Modification of Eagle's Medium with 20 mM HEPES and 4,5 g Dextrose/liter, without L-glutamine or Sodium bicarbonate is ideally suited for supporting and maintaining a vast range of mammalian cell types.
- Most widely used modification of Eagle's Basal Medium (BME)
- Contains four times greater concentration of amino acids, vitamins and supplementary components
- For optimal culturing of other cell types, a modification of 4500 mg/L glucose is available
The original formulation calls for 1000 mg/L of glucose, which was first employed to support the polyoma virus in primary and secondary embryonic mouse cultures.