Human Serum, Pooled, MP Biomedicals
Catalog # ICNA092930949
Supplier: MP Biomedicals
Specifications
- Description:Human Type AB serum
- Origin:Human (Male Donors)
- Pk:100 mL
Specifications
About this item
Human pooled serum is used as a media component to neutralize cell free virus. It's used as a reagent in liposome binding assay and in preparation of sperm.
- Obtained from normal clotted blood
- Liquid media
- Store at —20 °C
- Media component for cell free virus, liposome binding assay, preparation of sperm
Each donor unit of plasma used in the preparation of this material was tested and found negative for HBsAg, HIV 1/2, anti-HCV, HCV-NAT, HIV-1 NAT, and syphilis by FDA regulation.