Anti-CX3CR1 Rabbit Polyclonal Antibody
NOVUNBP1-76949
- Antibody type:Primary
- Antigen name:Chemokine (C-X3-C motif) receptor 1
- Antigen symbol:CX3CR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:1524
- Antigen synonyms:GPR13G-protein coupled receptor 13|Beta chemokine receptor-like 1|CMKBRL1|CCRL1|G protein-coupled receptor 13|CMKDR1|CX3C chemokine receptor 1|Fractalkine receptor|CMK-BRL1|chemokine (C-C) receptor-like 1|V28C-X3-C CKR-1|chemokine (C-X3-C motif) receptor 1|chemokine (C-X3-C) receptor 1|CMK-BRL-1|GPRV28
- Storage buffer:PBS
- Storage temperature:Store at 4 °C short term. Aliquot and store at ‒20 °C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Antibody was raised against a peptide corresponding to amino acids 2 to 21 of human CX3CR1. The sequence differs from those of mouse and rat CX3CR1 by four amino acids (NP_001328).
- Purification:Immunogen affinity purified
- Pk:0,1 mg
The CX3CR1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CX3CR1. This antibody reacts with human, mouse, rat. The CX3CR1 Antibody has been validated for the following applications: Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 2 µg/ml
Type: Primary
Antigen: CX3CR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat