Order Entry
Czech Republic
ContactUsLinkComponent
Anti-PSMA/FOLH1/NAALADase I Mouse Monoclonal Antibody [clone: GCP-04]
Anti-PSMA/FOLH1/NAALADase I Mouse Monoclonal Antibody [clone: GCP-04]
Catalog # NOVUNBP1-45057
Supplier:  Novus Biologicals
Anti-PSMA/FOLH1/NAALADase I Mouse Monoclonal Antibody [clone: GCP-04]
Catalog # NOVUNBP1-45057
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Folate Hydrolase 1
  • Antigen symbol:
    PSMA/FOLH1/NAALADase I
  • Clonality:
    Monoclonal
  • Clone:
    GCP-04
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Pig,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Gene ID:
    2346
  • Antigen synonyms:
    glutamate carboxylase II|EC 3.4.17.21|GCPIImGCP|folate hydrolase (prostate-specific membrane antigen) 1|Prostate-specific membrane antigen|Folylpoly-gamma-glutamate carboxypeptidase|Membrane glutamate carboxypeptidase|GCP2|Cell growth-inhibiting gene 27 protein|PSMPteroylpoly-gamma-glutamate carboxypeptidase|Glutamate carboxypeptidase II|FOLHNAALADase I|NAALAdase|PSMAFGCP|N-acetylated alpha-linked acidic dipeptidase 1|NAALAD1N-acetylated-alpha-linked acidic dipeptidase I|cell growth-inhibiting protein 27|Folate hydrolase 1|prostate specific membrane antigen variant F|glutamate carboxypeptidase 2
  • Storage buffer:
    PBS (pH 7,4)
  • Storage temperature:
    Store at 4 °C. Do not freeze.
  • Concentration:
    1,0 mg/ml
  • Immunogen:
    Recombinant fragment of human GCPII (amino acids 44-750) produced in S2 cells (NM_001014986.1).
  • Purification:
    Protein A purified
  • Pk:
    100 µG

Specifications

About this item

The PSMA / FOLH1 / NAALADase I Antibody (GCP-04) from Novus Biologicals is a mouse monoclonal antibody to PSMA / FOLH1 / NAALADase I. This antibody reacts with human, mouse, rat, porcine. The PSMA / FOLH1 / NAALADase I Antibody (GCP-04) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: PSMA/FOLH1/NAALADase I
Clonality: Monoclonal
Clone: GCP-04
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Porcine