Order Entry
Czech Republic
ContactUsLinkComponent
Anti-TNF-alpha Rabbit Polyclonal Antibody
Anti-TNF-alpha Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-19532
Supplier:  Novus Biologicals
Anti-TNF-alpha Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-19532
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tumor Necrosis Factor Alpha
  • Antigen symbol:
    TNF-alpha
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    7124
  • Antigen synonyms:
    Tumor necrosis factor ligand superfamily member 2|tumor necrosis factor alpha|member 2)|DIF|tumor necrosis factor|monocyte-derived|TNFATNF|tumor necrosis factor (TNF superfamily|member 2|TNFSF2TNF superfamily|TNF-a|macrophage-derived|TNF|TNF-alphacachectin|Cachectin|tumor necrosis factor-alpha|APC1 protein
  • Storage buffer:
    PBS
  • Storage temperature:
    Store at 4 °C short term. Aliquot and store at ‒20 °C long term. Avoid freeze-thaw cycles.
  • Concentration:
    1,0 mg/ml
  • Immunogen:
    A synthetic peptide made to an internal portion of the human TNF alpha protein (between residues 100-200) [UniProt P01375]
  • Purification:
    Immunogen affinity purified
  • Pk:
    100 µG

Specifications

About this item

The TNF-alpha Antibody from Novus Biologicals is a rabbit polyclonal antibody to TNF-alpha. This antibody reacts with human, mouse, rat. The TNF-alpha Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:50 to 1:200

Type: Primary
Antigen: TNF-alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat