Anti-CTR9 Rabbit Polyclonal Antibody
Catalog # NOVUNB100-68205
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:CTR9 homolog, Paf1/RNA polymerase II complex component
- Antigen symbol:CTR9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:9646
- Antigen synonyms:KIAA0155SH2 domain-binding protein 1|SH2-binding phosphoprotein|homolog (S. cerevisiae)|p150TSP|SH2 domain binding protein 1 (tetratricopeptide repeat containing)|RNA polymerase-associated protein CTR9 homolog|Paf1/RNA polymerase II complex component|SH2BP1p150|Ctr9|TSBP|TPR-containing
- Storage buffer:Tris-Citrate/Phosphate (pH 7,0 - 8,0)
- Storage temperature:Store at 4 °C. Do not freeze.
- Concentration:1 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 1123 and 1173 of human Paf1/RNA polymerase II complex component homolog CTR9 using the numbering given in entry NP_055448.1 (GeneID 9646).
- Purification:Immunogen affinity purified
- Pk:100 µl
Specifications
About this item
The CTR9 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CTR9. This antibody reacts with human, mouse. The CTR9 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:250
Type: Primary
Antigen: CTR9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse