Human AFP/Alpha 1 Fetoprotein PicoKine; ELISA Kit
Catalog # BSBTEK0651
Supplier: Boster Bio
CAS Number:
Specifications
- Pk:1 KIT
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Description:Human AFP/Alpha 1 Fetoprotein PicoKine; ELISA kit, 96 wells/kit
Specifications
About this item
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Human AFP.
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
Detect Human Alpha-Fetoprotein/AFP with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Alpha-Fetoprotein/AFP tissue specificity: Plasma. Synthesised by the fetal liver and yolk sac.
96wells/kit, with removable strips.