PicoKine Human Relaxin 2; ELISA Kit
Katalogové číslo BSBTEK1428
Supplier: Boster Bio
CAS Number:
Specifications
- Pk:1 KIT
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Description:Human Relaxin 2 PicoKine; ELISA kit
- Tests per kit:96 wells
Specifications
About this item
Sandwich high sensitivity ELISA kit for quantitative detection of human Relaxin 2.
- Sensitivity, detect single digit picogram level of target protein
- Specificity, thoroughly tested for cross-reactivity with family members
- Stability, guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, on-demand technical support directly from ELISA experts
Detect Human Relaxin-2/RLN2 with <4 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Relaxin-2/RLN2 tissue specificity: Isoform 1 is expressed in the ovary during pregnancy. Also expressed in placenta, decidua and prostate. Isoform 2 is relatively abundant in placenta. It is in much lower abundance in the prostate gland. Not detected in the ovary. .
96wells/kit, with removable strips.