Order Entry
Czech Republic
ContactUsLinkComponent
Platelet Activating Factor (PAF) ELISA Kit
Platelet Activating Factor (PAF) ELISA Kit
Catalog # ABCAAB287801-96
Supplier:  Abcam
CAS Number:  
undefined
Platelet Activating Factor (PAF) ELISA Kit
Catalog # ABCAAB287801-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Competitive
  • Host:
  • Primary antibody reactivity:
  • Description:
    Platelet activating factor ELISA kit
  • Environmentally Preferable:
  • Tests per kit:
    100 assays

Specifications

About this item

A sandwich ELISA kit for the quantitative measurement of platelet activating factor in bio fluids.

  • Assay Precision: Intra-Assay: CV <8%; Inter-Assay: CV <10%
  • Detection Range: 0,156 to 10 ng/ml
  • Sensitivity: <0,094 ng/ml
  • Easy, convenient and time-saving method to measure the level of PAF in serum, plasma, tissue homogenates, culture supernatants and other biological fluids
  • No significant cross-reactivity or interference was observed

Platelet-activating factor (PAF) is a potent phospholipid activator and mediator of many leukocyte functions, platelet aggregation and degranulation, inflammation, and anaphylaxis. It is also involved in changes to vascular permeability, the oxidative burst, chemotaxis of leukocytes, as well as augmentation of arachidonic acid metabolism in phagocytes. BioVision’s PAF ELISA kit is based on Competitive ELISA assay for the quantitative measurement of PAF in serum, plasma, and cell culture supernatants. The intensity of the colour is inversely proportional to the amount of PAF captured from the samples.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.