Cyclic di-GMP (sodium salt) ≥95%
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:2222132-40-1
:
Formula:C₂₀H₂₂N₁₀O₁₄P₂ ·2Na
:734.4 g/mol
Storage Temperature:Freezer
Specification Test Results
- Formulation:A solid
- Purity:≥95%
- Stability:≥ 2 years
Specification Test Results
Frequently Bought Together